4,4'-(ɑ-Methylbenzylidene)bisphenol 99%

BD91879-5G BD91879-25G
BLDPBD91879-5GEA 7.4 EUR
BLDPBD91879-5G BLDPBD91879-25G
4,4'-(ɑ-Methylbenzylidene)bisphenol 99%
4,4'-(alpha-Methylbenzylidene)bisphenol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₂₀H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00134687
CAS Number: 1571-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR