4,4'-Methylenedi-o-toluidine 98%

BD92079-5G BD92079-10G
BLDPBD92079-5GEA 23.1 EUR
BLDPBD92079-5G BLDPBD92079-10G
4,4'-Methylenedi-o-toluidine 98%
4,4'-Methylenedi-o-toluidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₅H₁₈N₂
MW: 226.3200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00126963
CAS Number: 838-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR