4,4'-Dihydroxydiphenylmethane 98%

BD92170-5G BD92170-25G
BLDPBD92170-5GEA 17.9 EUR
BLDPBD92170-5G BLDPBD92170-25G
4,4'-Dihydroxydiphenylmethane 98%
4,4'-Dihydroxydiphenylmethane

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00002385
CAS Number: 620-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR