4,4'-Dipyridyl disulphide 98%

BD92294-10G BD92294-250MG
BLDPBD92294-10GEA 208 EUR
BLDPBD92294-10G BLDPBD92294-250MG
4,4'-Dipyridyl disulphide 98%
4,4'-Dipyridyl disulphide

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₈N₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00006423
CAS Number: 2645-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR