4,4'-Oxybis(benzoyl chloride) 97%

BD92340-250MG BD92340-1G
BLDPBD92340-250MGEA 6.3 EUR
BLDPBD92340-250MG BLDPBD92340-1G
4,4'-Oxybis(benzoyl chloride) 97%
4,4'-Oxybis(benzoyl chloride)

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₄H₈Cl₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00496619
CAS Number: 7158-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR