Fmoc-Lys(Alloc)-OH 98%

BD9243-10G BD9243-25G
BLDPBD9243-10GEA 44.1 EUR
BLDPBD9243-10G BLDPBD9243-25G
Fmoc-Lys(Alloc)-OH 98%
Fmoc-Lys(Alloc)-OH

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₂₅H₂₈N₂O₆
MW: 452.507 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190872
CAS Number: 146982-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR