4,6-Dimethylindoline-2,3-dione 95%

BD92865-250MG BD92865-1G
BLDPBD92865-250MGEA 68.3 EUR
BLDPBD92865-250MG BLDPBD92865-1G
4,6-Dimethylindoline-2,3-dione 95%
4,6-Dimethylindoline-2,3-dione

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD03103379
CAS Number: 49820-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR