4-Amino-2-chloropyrimidine 98%

BD9409-5G BD9409-25G
BLDPBD9409-5GEA 13.7 EUR
BLDPBD9409-5G BLDPBD9409-25G
4-Amino-2-chloropyrimidine 98%
4-Amino-2-chloropyrimidine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₄H₄ClN₃
Storage Temperature: Ambient
MDL Number: MFCD00194051
CAS Number: 7461-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR