(4-Bromo-2-fluorophenyl)acetonitrile 98%

BD94875-10G BD94875-25G
BLDPBD94875-10GEA 27.3 EUR
BLDPBD94875-10G BLDPBD94875-25G
(4-Bromo-2-fluorophenyl)acetonitrile 98%
(4-Bromo-2-fluorophenyl)acetonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD07368653
CAS Number: 114897-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR