4-Bromo-o-anisaldehyde 98%

BD94900-1G BD94900-10G
BLDPBD94900-1GEA 9.5 EUR
BLDPBD94900-1G BLDPBD94900-10G
4-Bromo-o-anisaldehyde 98%
4-Bromo-o-anisaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD08276823
CAS Number: 43192-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR