4-Bromo-2-methoxypyridine 98%

BD94904-1G BD94904-100G
BLDPBD94904-1GEA 8.4 EUR
BLDPBD94904-1G BLDPBD94904-100G
4-Bromo-2-methoxypyridine 98%
4-Bromo-2-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₆BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD08277268
CAS Number: 100367-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR