4'-Bromo-3'-fluoroacetanilide 95%

BD95014-25G BD95014-1G
BLDPBD95014-25GEA 210 EUR
BLDPBD95014-25G BLDPBD95014-1G
4'-Bromo-3'-fluoroacetanilide 95%
4'-Bromo-3'-fluoroacetanilide

Storage condition: Sealed in dry, Room Temperature

Formula: H₃CC(O)NH(C₆H₃BrF)
Storage Temperature: Ambient
MDL Number: MFCD00672937
CAS Number: 351-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR