4-Bromoisoindoline hydrochloride 95%

BD95249-100MG BD95249-10G
BLDPBD95249-100MGEA 20 EUR
BLDPBD95249-100MG BLDPBD95249-10G
4-Bromoisoindoline hydrochloride 95%
4-Bromoisoindoline hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₉BrClN
MW: 234.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10700134
CAS Number: 923590-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR