4-Bromothiophene-2-carbaldehyde 98%

BD9527-10G BD9527-100G
BLDPBD9527-10GEA 13.7 EUR
BLDPBD9527-10G BLDPBD9527-100G
4-Bromothiophene-2-carbaldehyde 98%
4-Bromothiophene-2-carbaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₃BrOS
MW: 191.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005431
CAS Number: 18791-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR