4-Bromo-m-xylene 98%
|
|
BD95279-100G
BD95279-25G
BLDPBD95279-100GEA
74.6
EUR
BLDPBD95279-100G
BLDPBD95279-25G
4-Bromo-m-xylene 98%
4-Bromo-m-xylene
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 214 Melting Pt: -1 Storage Temperature: Ambient |
MDL Number:
MFCD00000071 CAS Number: 583-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...