(4-Bromopyridine-2,6-diyl)dimethanol 95%
|
|
BD95361-100MG
BD95361-1G
BLDPBD95361-100MGEA
94.5
EUR
BLDPBD95361-100MG
BLDPBD95361-1G
(4-Bromopyridine-2,6-diyl)dimethanol 95%
(4-Bromopyridine-2,6-diyl)dimethanol
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₈BrNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09910217 CAS Number: 120491-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...