2-Bromo-5-methylthiophene 98% stabilised


Stabilization: Stabilised with copper chip + NaHCO₃

BD9568-5G BD9568-100G
BLDPBD9568-5GEA 8.4 EUR
BLDPBD9568-5G BLDPBD9568-100G
2-Bromo-5-methylthiophene 98% stabilised
2-Bromo-5-methylthiophene

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₅BrS
Storage Temperature: Refrigerator
MDL Number: MFCD00130103
CAS Number: 765-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR