4-Chloro-2-nitropyridine 97%

BD95820-10G BD95820-100G
BLDPBD95820-10GEA 110 EUR
BLDPBD95820-10G BLDPBD95820-100G
4-Chloro-2-nitropyridine 97%
4-Chloro-2-nitropyridine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114167
CAS Number: 65370-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR