3,5-Dimethylisoxazole-4-sulphonyl chloride 98%

BD9591-25G BD9591-1G
BLDPBD9591-25GEA 83 EUR
BLDPBD9591-25G BLDPBD9591-1G
3,5-Dimethylisoxazole-4-sulphonyl chloride 98%
3,5-Dimethylisoxazole-4-sulphonyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₆ClNO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00052554
CAS Number: 80466-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR