Thymine 97%

BD9599-5G BD9599-1000G
BLDPBD9599-5GEA 6.3 EUR
BLDPBD9599-5G BLDPBD9599-1000G
Thymine 97%
Thymine

Storage condition: Sealed in dry, Room Temperature

Formula: C₅H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00006026
CAS Number: 65-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR