4-Chloro-N-methoxy-N-methylbenzamide 97%

BD96407-1G BD96407-100G
BLDPBD96407-1GEA 8.4 EUR
BLDPBD96407-1G BLDPBD96407-100G
4-Chloro-N-methoxy-N-methylbenzamide 97%
4-Chloro-N-methoxy-N-methylbenzamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD02684308
CAS Number: 122334-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR