4-Chloropicolinaldehyde 97%

BD96480-100G BD96480-25G
BLDPBD96480-100GEA 774 EUR
BLDPBD96480-100G BLDPBD96480-25G
4-Chloropicolinaldehyde 97%
4-Chloropicolinaldehyde

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₆H₄ClNO
Storage Temperature: Freezer
MDL Number: MFCD07437896
CAS Number: 63071-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR