4-Ethynylpyridine hydrochloride 97%
|
|
BD97056-250MG
BD97056-10G
BLDPBD97056-250MGEA
10.5
EUR
BLDPBD97056-250MG
BLDPBD97056-10G
4-Ethynylpyridine hydrochloride 97%
4-Ethynylpyridine hydrochloride
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₆ClN Storage Temperature: Refrigerator |
MDL Number:
MFCD03093912 CAS Number: 352530-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...