4-Fluoro-N-methylbenzenesulphonamide 98%

BD97348-1G BD97348-25G
BLDPBD97348-1GEA 50.4 EUR
BLDPBD97348-1G BLDPBD97348-25G
4-Fluoro-N-methylbenzenesulphonamide 98%
4-Fluoro-N-methylbenzenesulphonamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₈FNO₂S
Storage Temperature: Ambient
MDL Number: MFCD01211989
CAS Number: 433-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR