4-Hexen-1-ol 97%

BD97492-5G BD97492-25G
BLDPBD97492-5GEA 49.4 EUR
BLDPBD97492-5G BLDPBD97492-25G
4-Hexen-1-ol 97%
4-Hexen-1-ol

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₁₂O
MW: 100.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009713
CAS Number: 6126-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR