4-Methoxyphenyl-β-D-galactopyranoside 98%

BD98582-1G BD98582-25G
BLDPBD98582-1GEA 42 EUR
BLDPBD98582-1G BLDPBD98582-25G
4-Methoxyphenyl-β-D-galactopyranoside 98%
4-Methoxyphenyl-beta-D-galactopyranoside

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₁₃H₁₈O₇
MW: 286.28 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06797126
CAS Number: 3150-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR