4-Methyl-4,5,6,7-tetrahydrothieno[3,2-c]pyridine 97%

BD98886-250MG BD98886-1G
BLDPBD98886-250MGEA 111 EUR
BLDPBD98886-250MG BLDPBD98886-1G
4-Methyl-4,5,6,7-tetrahydrothieno[3,2-c]pyridine 97%
4-Methyl-4,5,6,7-tetrahydrothieno[3,2-c]pyridine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₈H₁₁NS
Storage Temperature: Refrigerator
MDL Number: MFCD07366382
CAS Number: 92503-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR