2-Amino-5-fluorophenol 98%

BD9964-5G BD9964-500G
BLDPBD9964-5GEA 14.7 EUR
BLDPBD9964-5G BLDPBD9964-500G
2-Amino-5-fluorophenol 98%
2-Amino-5-fluorophenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₆FNO
MW: 127.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00671759
CAS Number: 53981-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR