VWR®, Instrument Calibration Standard 1, 4 components: 5000 mg/l each of Ca, K, Mg, Na in 5% HNO₃
91176.180EA
204
EUR
91176.180
91176.260
VWR®, Instrument Calibration Standard 1, 4 components: 5000 mg/l each of Ca, K, Mg, Na in 5% HNO₃
Standards
Cationic Standards
Multi-Element Standards
Certifications: ISO 17025/17034