VWR®, Internal Standard Mix, 7 components: 10 ug/ml each of 6Li, Sc, Ge, Y, in, Tb, Bi in 5% HNO₃
91178.260EA 444 EUR
91178.260 91178.180
VWR®, Internal Standard Mix, 7 components: 10 ug/ml each of 6Li, Sc, Ge, Y, in, Tb, Bi in 5% HNO₃
Standards Cationic Standards Multi-Element Standards
Certifications: ISO 17025/17034