VWR®, Interference Check Solution 1, 12 components in Nitric Acid 5%, Hydrofluoric acid tr%
91189.260EA 730 EUR
91189.260 91189.180
VWR®, Interference Check Solution 1, 12 components in Nitric Acid 5%, Hydrofluoric acid tr%
Standards Cationic Standards Multi-Element Standards
12 components: Chlorides (Cl-) 10000 mg/l, Carbon (C) 2000 mg/l, Aluminium (Al) 1000 mg/l, Calcium (Ca) 1000 mg/l, Iron (Fe) 1000 mg/l, Potassium (K) 1000 mg/l, Magnesium (Mg) 1000 mg/l, Sodium (Na) 1000 mg/l, Phosphorus (P) 1000 mg/l, Sulphur (S) 1000 mg/l, Molybdenum (Mo) 20 mg/l, Titanium (Ti) 20 mg/l in 5% HNO₃/tr HF
Certifications: ISO 17025/17034