VWR®, ICP-OES Wavelength calibration solution, 15 components in 5% HNO₃
91216.260EA 586 EUR
91216.260 91216.180
VWR®, ICP-OES Wavelength calibration solution, 15 components in 5% HNO₃
Standards Cationic Standards Multi-Element Standards
15 components: Al 5 ug/ml, As 5 ug/ml, Ba 5 ug/ml, Cd 5 ug/ml, Co 5 ug/ml, Cr 5 ug/ml, Cu 5 ug/ml, Mn 5 ug/ml, Mo 5 ug/ml, Ni 5 ug/ml, Pb 5 ug/ml, Se 5 ug/ml, Sr 5 ug/ml, Zn 5 ug/ml, K 50 ug/ml in 5% HNO₃
Certifications: ISO 17025/17034