VWR®, Combined Six Cation Standard-I, 6 components in H₂O
91290.154EA 133 EUR
91290.154 91290.180
VWR®, Combined Six Cation Standard-I, 6 components in H₂O
Standards Cationic Standards Multi-Element Standards
6 components: Lithium (Li+) 50 mg/l, Sodium (Na+) 200 mg/l, Ammonium (NH4+) 400 mg/l, Potassium (K+) 200 mg/l, Calcium (Ca2+) 1000 mg/l, Magnesium (Mg2+) 200 mg/l in H₂O
Certifications: ISO 17025/17034