Laboratory Freezers, ZLN 85
Supplier: POL-EKO APARATURA SP.
ZLN85CSMART ZLN85CSSMART ZLN85PSMART ZLN85PSSMART ZLN85PSMPR ZLN85PSSMPR
POLEZLN85CSMARTEA 2050 EUR
POLEZLN85CSMART POLEZLN85CSSMART POLEZLN85PSMART POLEZLN85PSSMART POLEZLN85PSMPR POLEZLN85PSSMPR
Laboratory Freezers, ZLN 85
Freezers Laboratory / General Purpose Freezers
Laboratory freezers ZLN 85 temperature range of −25 to 0 °C and a user-friendly interface. Conduct precise experiments with quality control at −20 °C, and multilingual support. Stay connected with LAN and USB ports, enhance security with door locks and alarms. The Smart Pro edition boasts WiFi communication and LabDesk software.
  • Quality control protocol (at −20 °C)
  • Instruction manual in English and menu in various languages
  • Open door alarm
  • LAN port and USB port
  • Access port (Ø 20 mm, on the left side) for an external sensor
  • Door lock
  • Full external doors
More Product Information