Ultra-Low Freezers, ZLN-UT 500 VIP
Supplier: POL-EKO APARATURA SP.
ZLNUT500VIPCSM ZLNUT500VIPPSP ZLNUT500VIPPSM
POLEZLNUT500VIPCSMEA 10350 EUR
POLEZLNUT500VIPCSM POLEZLNUT500VIPPSP POLEZLNUT500VIPPSM
Ultra-Low Freezers, ZLN-UT 500 VIP
Freezers Ultra Low Temperature Freezers
Revolutionise your freezing experience with ZLN-UT Ultra-Low Freezers, reaching an impressive ‒86 °C. Manufactured with VIP insulated panels, these freezers ensure long-term preservation for biotechnological samples. These 'As Smart As Your Smartphone' ultra-low freezers offer reliability and flexibility. Choice of either COMFORT model with a stainless steel interior, which is perfect for general use or the PREMIUM model, featuring an acid-resistant stainless steel interior, which is ideal for specialised applications.
  • Quality control protocol (at ‒80 °C)
  • Instruction manual in English and menu in various languages
  • Open door alarm with door lock
  • Units have castors as standard
  • LAN port and USB port
  • Full solid shelves with a central hole
  • Solid external doors and each inner compartment has a solid door
  • WiFi communication and LAN cable (Smart Pro only)
  • LabDesk software for, amongst other things, downloading data to computer (only Smart Pro, optional for Smart version)
These freezers provide a temperature range from ‒86 to ‒50 °C, ensuring optimal conditions for diverse applications. Equipped with essential features such as a quality control protocol (at ‒80 °C), multilingual instruction manuals, and a user-friendly menu, these freezers prioritise precision and ease of use. The open-door alarm adds an extra layer of security, alerting users to potential temperature fluctuations. The inclusion of castors enhances mobility, making these freezers adaptable to various laboratory setups. The environmentally friendly refrigerant (R290) has a very low global warming potential (GWP is 3).
For seamless connectivity, ZLN-UT 500 VIP Ultra-Low Freezers feature LAN and USB ports, allowing easy integration into your laboratory network. The left-side access port accommodates an external sensor (Ø20 mm), providing additional monitoring capabilities. The door lock ensures the safety and integrity of valuable samples.
The Smart Pro edition provides the next level of convenience with WiFi communication. This edition also includes a LAN cable, facilitating high-speed data transfer and communication within the laboratory setting. For advanced data management, the LabDesk software, available exclusively in the Smart Pro version (and optional for the Smart version), allows you to download and organise freezer data directly to your computer.
With full shelves featuring holes for efficient storage, full external doors ensuring easy accessibility, and individual full doors for each internal chamber, ZLN-UT 500 VIP Ultra-Low Freezers combine functionality with a sleek design. Elevate your laboratory freezing experience with these state-of-the-art freezers, providing unmatched performance and reliability.
Unit has two compartments which each accommodate 4+8 racks, 12/16 boxes per rack, 176 boxes per compartment, giving a total of 352 boxes per unit. 28512 × test tubes (Ø 12,5 mm). Natural air convection with two internal chambers and with powder coated sheet housing.
Ordering information: Accessories are not included and must be ordered separately.
Delivery information: User manual can be downloaded from controller, quick start guide supplied in chamber.