Cooled Incubators, ST 2
Supplier: POL-EKO APARATURA SP.
ST2CSMART ST2CSSMART ST2PSMART ST2PSSMART ST2PSMARTPRO ST2PSSMARTPRO
POLEST2CSMARTEA 1820 EUR
POLEST2CSMART POLEST2CSSMART POLEST2PSMART POLEST2PSSMART POLEST2PSMARTPRO POLEST2PSSMARTPRO
Cooled Incubators, ST 2
Incubators Cooled Incubators
ST 2 Cooled Incubators ensure reliable temperature control from 3 to 70 °C, regardless of ambient conditions. POL-EKO's models boast internal LED lighting, secure lockable doors. Ideal for stability testing and pharmaceutical storage, these incubators offer versatile chamber configurations, optimising laboratory space and enabling independent temperature control in each compartment.
  • Quality control protocol (at 37 °C)
  • Instruction manual in English and menu in various languages
  • Over temperature protection 1.0 class (according to DIN 12880) for C (comfort) models, 2.0 for P (premium) models and 3.3 class in Smart PRO
  • Open door alarm; Door lock; Solid external doors
  • LAN port and USB port; Access port (Ø30 mm, on the left side) for an external sensor
  • Internal LED light
  • Stainless steel wire shelves for models C (comfort) and P (premium)
  • Wi-Fi communication and LAN cable (Smart Pro only)
  • LabDesk software for, amongst other things, downloading data to computer (Smart Pro only, optional for Smart version)
Experience unparalleled precision in sample incubation with POL-EKO's ST 2 Cooled Incubators. These innovative thermostatic devices, available in comfort (C) and premium (P) models, ensure reliable temperature control from 3 to 70 °C. Equipped with both heating and cooling systems, these incubators guarantee optimal conditions for storage and incubation above or below ambient temperature.
The Smart and Smart PRO controller versions provide advanced features as standard, including over-temperature protection, open door alarms, access ports for external sensors, LAN and USB ports, and internal LED lighting. The user-friendly design includes a lockable door, stainless steel wire shelves (INOX), and solid external doors for convenience and security. With a chamber capacity of 150 L and working capacity of 122 L, these incubators offer ample space for various applications. The optional LabDesk software for Smart Pro models, facilitates data download to computers.
Choose the Smart or Smart PRO controller based on your specific requirements. Upgrade your laboratory capabilities with POL-EKO's Cooled Incubators ST 2, setting new standards in temperature control and reliability. The environmentally friendly refrigerant (R1234ze) has a very low global warming potential (GWP is 1).
Designed for versatility, the ST 2 Cooled Incubators are also available in 2-chamber versions (models ST 2/2, ST 2/3), allowing independent control of each chamber. This innovative solution optimises laboratory space and enables testing at two different temperatures simultaneously. For added flexibility, multi-chamber devices are available in mixed versions, combining a thermostatic cabinet with a laboratory refrigerator or freezer, each with independent control. Please ask your Avantor sales representative for more information.
Delivery information: Supplied with 3 stainless steel wire shelves. Other accessories are not included and must be ordered separately. User manual can be downloaded from controller, quick start guide supplied in chamber.