Cooled Incubators, ST 4
Supplier: POL-EKO APARATURA SP.
ST4CSMART ST4CSSMART ST4PSMART ST4PSSMART ST4PSMARTPRO ST4PSSMARTPRO
POLEST4CSMARTEA 2090 EUR
POLEST4CSMART POLEST4CSSMART POLEST4PSMART POLEST4PSSMART POLEST4PSMARTPRO POLEST4PSSMARTPRO
Cooled Incubators, ST 4
Incubators Cooled Incubators
ST 4 Cooled Incubators ensure reliable temperature control from 3 to 70 °C, regardless of ambient conditions. POL-EKO's models boast internal LED lighting, secure lockable doors. Ideal for stability testing and pharmaceutical storage, these incubators offer versatile chamber configurations, optimising laboratory space and enabling independent temperature control in each compartment.
  • Quality control protocol (at 37 °C)
  • Instruction manual in English and menu in various languages
  • Over temperature protection 1.0 class (according to DIN 12880) for C (comfort) models, 2.0 for P (premium) models and 3.3 class in Smart PRO
  • Open door alarm; Door lock; Solid external doors
  • LAN port and USB port; Access port (Ø30 mm, on the left side) for an external sensor
  • Internal LED light
  • Stainless steel wire shelves for C (comfort) and P (premium) models
  • Wi-Fi communication and LAN cable (Smart Pro only)
  • LabDesk software for, amongst other things, downloading data to computer (Smart Pro only, this is optional for Smart version)
Experience unparalleled precision in sample incubation with POL-EKO's ST 4 Cooled Incubators. These innovative thermostatic devices, available in comfort (C) and premium (P) models, ensure reliable temperature control from 3 to 70 °C. Equipped with both heating and cooling systems, these incubators guarantee optimal conditions for storage and incubation above or below ambient temperature.
The Smart and Smart PRO controller versions provide advanced features as standard, including over-temperature protection, open door alarms, access ports for external sensors, LAN and USB ports, and internal LED lighting. The user-friendly design includes a lockable door, stainless steel wire shelves (INOX), and solid external doors for convenience and security. With a chamber capacity of 250 L and working capacity of 203 L, these incubators offer ample space for various applications. The optional LabDesk software for Smart Pro models, facilitates data download to computers.
Choose the Smart or Smart PRO controller based on your specific requirements. Upgrade your laboratory capabilities with POL-EKO's ST 4 Cooled Incubators, setting new standards in temperature control and reliability. The environmentally friendly refrigerant (R1234ze) has a very low global warming potential (GWP is 1).
Delivery information: Supplied with 4 stainless steel wire shelves. Other accessories are not included and must be ordered separately. User manual can be downloaded from controller, quick start guide supplied in chamber.