Cooled Incubators, ST 500
Supplier: POL-EKO APARATURA SP.
ST500CMSMART ST500CMSSMART ST500PMSMART ST500PMSSMART ST500PMSMPRO ST500PMSSMPRO
POLEST500CMSMARTEA 3760 EUR
POLEST500CMSMART POLEST500CMSSMART POLEST500PMSMART POLEST500PMSSMART POLEST500PMSMPRO POLEST500PMSSMPRO
Cooled Incubators, ST 500
Incubators Cooled Incubators
ST 500 Cooled incubators ensure reliable temperature control from 3 to 70 °C, regardless of ambient conditions. POL-EKO's models boast internal LED lighting, secure lockable doors. Ideal for stability testing and pharmaceutical storage, these incubators offer versatile chamber configurations, optimising laboratory space and enabling independent temperature control in each compartment.
  • Quality control protocol (at 37 °C)
  • Instruction manual in English and menu in various languages
  • Over temperature protection 1.0 class (according to DIN 12880) for C (comfort) models, 2.0 for P (premium) models and 3.3 class in Smart PRO
  • Assembly kit for fixing to the wall; Stainless steel wire shelves for C (comfort) and P (premium) models
  • Monoblock cooling unit; Open door alarm; Door lock; Solid external doors
  • LAN port and USB port; WiFi communication (Smart Pro only); LAN cable (Smart Pro only)
  • Internal LED light; Access port (Ø 30 mm, on the left side) for an external sensor
  • LabDesk software for, amongst other things, downloading data to computer (Smart Pro only, this is optional for Smart version)
Elevate your laboratory capabilities with the ST 500 cooled incubator COMFORT or PREMIUM models, featuring a SMART/SMART PRO controller for precision. Providing a stabile temperature range from 3 to 40 °C and forced air convection, these incubators guarantee reliable sample storage and incubation. The 500 L chamber capacity, three stainless steel wire shelves, and a 30 mm access port provide ample space and flexibility.
Benefit from features like USB port, automatic defrosting, and interior LED lighting. The solid door, door lock, and monoblock cooling unit contribute to optimal performance. The 24-month warranty adds peace of mind. Explore the Smart Pro options for enhanced communication, including WiFi, LAN port, and LabDesk software.
Discover unparalleled control with the ST 500 cooled incubators. The solid door, stainless steel wire shelves, and access port enhance usability. Security features like over-temperature protection, open door alarm, and door lock guarantee a controlled environment. Benefit from additional features such as LAN and USB ports, and optional Smart Pro communication capabilities. Trust POL-EKO for reliable, efficient, and technologically advanced cooled incubators. The environmentally friendly refrigerant (R290) has a very low global warming potential (GWP is 3).
Delivery information: Supplied with 3 stainless steel wire shelves. Other accessories are not included and must be ordered separately. User manual can be downloaded from controller, quick start guide supplied in chamber.