Cooled Incubators, ST 1200
Supplier: POL-EKO APARATURA SP.
ST1200CMSMART ST1200CMSSMART ST1200PMSMART ST1200PMSMPRO ST1200PMSSMART ST1200PMSSMPRO
POLEST1200CMSMARTEA 5780 EUR
POLEST1200CMSMART POLEST1200CMSSMART POLEST1200PMSMART POLEST1200PMSMPRO POLEST1200PMSSMART POLEST1200PMSSMPRO
Cooled Incubators, ST 1200
Incubators Cooled Incubators
ST 1200 Cooled incubators ensure reliable temperature control from 3 to 70 °C, regardless of ambient conditions. POL-EKO's models boast internal LED lighting, secure lockable doors. Ideal for stability testing and pharmaceutical storage, these incubators offer versatile chamber configurations, optimising laboratory space and enabling independent temperature control in each compartment.
  • Quality control protocol (at 37 °C)
  • Instruction manual in English and menu in various languages
  • Over temperature protection 1.0 class (according to DIN 12880) for C (comfort) models, 2.0 for P (premium) models and 3.3 class in Smart PRO
  • Monoblock cooling unit; Castors; Open door alarm; Door lock; Solid double external doors
  • Assembly kit for fixing to the wall; Stainless steel wire shelves for C (comfort) and P (premium) models
  • LAN port and USB port; WiFi communication (Smart Pro only); LAN cable (Smart Pro only)
  • Internal LED light; Access port (Ø 30 mm, on the left side) for an external sensor
  • LabDesk software for, amongst other things, downloading data to computer (Smart Pro only, this is optional for Smart version)
Elevate your laboratory capabilities with the ST 1200 cooled incubator COMFORT or PREMIUM models, featuring a SMART/SMART PRO controller for precision. Providing a stabile temperature range from 3 to 40 °C and forced air convection, these incubators guarantee reliable sample storage and incubation. The 1365 L chamber capacity, three stainless steel wire shelves, and a 30 mm access port provide ample space and flexibility.
Benefit from features like USB port, automatic defrosting, and interior LED lighting. The solid door, door lock, and monoblock cooling unit contribute to optimal performance. The 24-month warranty adds peace of mind. Explore the Smart Pro options for enhanced communication, including WiFi, LAN port, and LabDesk software.
Discover unparalleled control with the cooled incubators ST 1200. The solid door, stainless steel wire shelves, and access port enhance usability. Security features like over-temperature protection, open door alarm, and door lock guarantee a controlled environment. Benefit from additional features such as LAN and USB ports, and optional Smart Pro communication capabilities. Trust POL-EKO for reliable, efficient, and technologically advanced cooled incubators. The environmentally friendly refrigerant (R290) has a very low global warming potential (GWP is 3).
Delivery information: Supplied with 2×3 stainless steel wire shelves. Other accessories are not included and must be ordered separately. User manual can be downloaded from controller, quick start guide supplied in chamber.