Cooled Incubators, ST 700 for FIT or FOT system
Supplier: POL-EKO APARATURA SP.
ST700CSMART ST700CSSMART ST700PSMART ST700PSSMART ST700PSMARTPRO ST700PSSMPRO
POLEST700CSMARTEA 0 EUR
POLEST700CSMART POLEST700CSSMART POLEST700PSMART POLEST700PSSMART POLEST700PSMARTPRO POLEST700PSSMPRO
Cooled Incubators, ST 700 for FIT or FOT system
Incubators Cooled Incubators
ST 700 Cooled incubators operate between 3 to 50 °C, these incubators offer internal LED lighting, secure lockable doors, and versatile chamber configurations. Ideal for stability testing. Explore the FOT system for day-night simulation in the COMFORT and PREMIUM models or the PREMIUM Smart PRO with FIT for advanced intensity control.
  • Quality control protocol (at 37 °C)
  • Instruction manual in English and menu in various languages
  • Over temperature protection 1.0 class (according to DIN 12880) for C (comfort) models, 2.0 for P (premium) models and 3.3 class in Smart PRO
  • Monoblock cooling unit; Open door alarm; Door lock; Solid external doors
  • Assembly kit for fixing to the wall; Stainless steel wire shelves for C (comfort) and P (premium) models
  • LAN port and USB port; WiFi communication (Smart Pro only); LAN cable (Smart Pro only)
  • Internal LED light; Access port (Ø30 mm, on the left side) for an external sensor
  • LabDesk software for, amongst other things, downloading data to computer (Smart Pro only, this is optional for Smart version)
Elevate your laboratory capabilities with the ST 700 cooled incubator COMFORT or PREMIUM models, featuring a SMART/SMART PRO controller for precision. Providing a stabile temperature range from 3 to 50 °C and forced air convection, these incubators guarantee reliable sample storage and incubation. The 625 L chamber capacity, three stainless steel wire shelves, and a 30 mm access port provide ample space and flexibility.
Benefit from features like USB port, automatic defrosting, and interior LED lighting. The solid door, door lock, and monoblock cooling unit contribute to optimal performance. The 24-month warranty adds peace of mind. Explore the Smart Pro options for enhanced communication, including WiFi, LAN port, and LabDesk software.
Discover unparalleled control with the cooled incubators ST 700. Security features like over-temperature protection, open door alarm, and door lock guarantee a controlled environment. Benefit from additional features such as LAN and USB ports, and optional Smart Pro communication capabilities.
In addition to the same functionality as the standard ST 700 models, the photoperiodic (FOT) systems for day and night simulation or the Phytotron (FIT) systems (either of these systems must be factory fitted) provide additional intensity control. Choose the COMFORT or PREMIUM Smart PRO version of these advanced cooled incubators to unlock these innovative features. Trust POL-EKO for reliable, efficient, and technologically advanced cooled incubators. The environmentally friendly refrigerant (R290) has a very low global warming potential (GWP is 3).
Delivery information: Supplied with 3 stainless steel wire shelves. FIT/FOT panels are not included, please ask your Avantor sales representative for more information. Other accessories are not included and must be ordered separately. User manual can be downloaded from controller, quick start guide supplied in chamber.
Caution: Note: The optional photoperiodic (FOT) systems for day and night simulation or the Phytotron (FIT) systems must be factory fitted, these can not be retrofitted.