Methyl chloroformate 99%

Supplier: ACROS ORGANICS
Danger

126530050 126535000 126531000 126530025
ACRO126530050EA 48 EUR
ACRO126530050 ACRO126535000 ACRO126531000 ACRO126530025
Methyl chloroformate 99%
Methyl chloroformate
Formula: C₂H₃ClO₂
MW: 94.5 g/mol
Boiling Pt: 70…72 °C (760 mmHg)
Melting Pt: –61 °C
Density: 1.223 g/cm³
Flash Pt: 5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000639
CAS Number: 79-22-1
EINECS: 201-187-3
UN: 1238
ADR: 6.1
Merck Index: 12,06123

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3860 to 1.3880 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR