2-Acetylthiazole ≥99%
Supplier: ALFA AESAR
|
Warning
|
A11266.03
A11266.06
A11266.03EA
75.3
EUR
A11266.03
A11266.06
2-Acetylthiazole ≥99%
2-Acetylthiazole
Purity: 99%
CAS No.: 24295-03-2
Molecular Formula: C5H5NOS
Formula Weight: 127.17
Flash Point: 78°(172°F)
Refractive Index: 1.5480
Odour: Characteristic
Boiling Point: 89-91°/12mm
Beilstein Reference: 109803
MDL No.: MFCD00005324
Formula:
C₅H₅NOS Boiling Pt: 89…91 °C (12 mmHg) Melting Pt: 65,5 °C Density: 1,227 Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005324 CAS Number: 24295-03-2 EINECS: 246-134-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...