4-Benzylbiphenyl ≥98%
A15392.06
A15392.14
A15392.06EA
69.4
EUR
A15392.06
A15392.14
4-Benzylbiphenyl ≥98%
4-Benzylbiphenyl
Purity: 98+%
CAS No.: 613-42-3
Molecular Formula: C19H16
Formula Weight: 244.34
Flash Point: 208°(406°F)
Melting Point: 84-87°
Boiling Point: 285-286°/110mm
Beilstein Reference: 2047066
MDL No.: MFCD00041831
Formula:
C₁₉H₁₆ Boiling Pt: 285…286 °C (110 mmHg) Melting Pt: 84…87 °C Density: 1,171 g/cm³ (0 °C) Flash Pt: 208 °C (406 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041831 CAS Number: 613-42-3 EINECS: 210-341-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...