Tricarballylic acid ≥98%

Supplier: ALFA AESAR
Warning

B22103.06 B22103.14
B22103.06EA 45.2 EUR
B22103.06 B22103.14
Tricarballylic acid ≥98%
Tricarballylic acid

Purity: 98% CAS No.: 99-14-9 Molecular Formula: C6H8O6 Formula Weight: 176.12 Melting Point: 157-162° Beilstein Reference: 1783567 Merck Reference: 14,9622 MDL No.: MFCD00002723

Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 266
Melting Pt: 157…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00002723
CAS Number: 99-14-9
EINECS: 202-733-3
Merck Index: 12,09752

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR