Ethylurea ≥98%

Supplier: ALFA AESAR

B24040.22 B24040.36 B24040.14
B24040.22EA 156 EUR
B24040.22 B24040.36 B24040.14
Ethylurea ≥98%
Ethylurea

Purity: 98% CAS No.: 625-52-5 Molecular Formula: CH3CH2NHCONH2 Formula Weight: 88.11 Melting Point: 88-94° Beilstein Reference: 1740512

Formula: C₃H₈N₂O
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 88…94 °C
Density: 1,213 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007953
CAS Number: 625-52-5
EINECS: 210-898-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR