4-Methylpyrocatechol 96%

Supplier: ALFA AESAR
Danger

L04495.14 L04495.22
L04495.14EA 33.6 EUR
L04495.14 L04495.22
4-Methylpyrocatechol 96%
4-Methylpyrocatechol

Purity: 96% CAS No.: 452-86-8 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 140°(284°F) Melting Point: 67-69° Boiling Point: 251° Beilstein Reference: 636512 MDL No.: MFCD00002205

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 251 °C
Melting Pt: 67…69 °C
Flash Pt: 140 °C (284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002205
CAS Number: 452-86-8
EINECS: 207-214-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR