cis-1,2-Cyclohexanedicarboxylic acid ≥98%

Warning

L10466.14 L10466.22
L10466.14EA 51.5 EUR
L10466.14 L10466.22
cis-1,2-Cyclohexanedicarboxylic acid ≥98%
cis-1,2-Cyclohexanedicarboxylic acid

Purity: 98% CAS No.: 610-09-3 Molecular Formula: C8H12O4 Formula Weight: 172.18 Melting Point: 189-192° Beilstein Reference: 2049754

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 339
Melting Pt: 189…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00064949
CAS Number: 610-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR