4-Bromo-2-fluorobenzyl alcohol ≥97%
Supplier: ALFA AESAR
|
Warning
|
L18679.06
L18679.03
L18679.06EA
67.5
EUR
L18679.06
L18679.03
4-Bromo-2-fluorobenzyl alcohol ≥97%
4-Bromo-2-fluorobenzyl alcohol
Purity: 97%
CAS No.: 188582-62-9
Molecular Formula: C7H6BrFO
Formula Weight: 205.03
Melting Point: 31-33°
Beilstein Reference: 7757667
Formula:
C₇H₆BrFO Melting Pt: 31…33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143265 CAS Number: 188582-62-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...