Piperidin-3-ol 98%

Danger

367080100 367080500
ACRO367080100EA 62.4 EUR
ACRO367080100 ACRO367080500
Piperidin-3-ol 98%
Piperidin-3-ol
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 67…69 °C (2 mmHg)
Melting Pt: 56…62 °C
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014591
CAS Number: 6859-99-0
EINECS: 229-957-4
UN: 3263
ADR: 8 II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder and/or lumps
Infrared spectrum Conforms
Melting point 56 to 62 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR