Piperonyl alcohol ≥98%
A12661.14
A12661.22
A12661.36
A12661.14EA
31.5
EUR
A12661.14
A12661.22
A12661.36
Piperonyl alcohol ≥98%
Piperonyl alcohol
Purity: 98%
CAS No.: 495-76-1
Molecular Formula: C8H8O3
Formula Weight: 152.15
Flash Point:
Formula:
C₈H₈O₃ Boiling Pt: 156…158 °C (16 mmHg) Melting Pt: 50…55 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005836 CAS Number: 495-76-1 EINECS: 207-808-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...